Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257831.1 | NP_001244760.1 | 493 | 116 | ||
XM_015152791.2 | XP_015008277.1 | 745 | 116 | |||
GenBank mRNA | JU335196.1 | - | 493 | 116 | ||
JU335197.1 | - | 493 | 116 | |||
JU470495.1 | - | 1643 | 116 | |||
JU470496.1 | - | 1599 | 116 | |||
JU470497.1 | - | 1647 | 116 | |||
JV043673.1 | - | 568 | 116 | |||
JV472877.1 | - | 390 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |