Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260980.1 | NP_001247909.1 | 252 | 73 | ||
GenBank mRNA | JU334909.1 | - | 251 | 73 | ||
JU475915.1 | - | 229 | 73 | |||
JV047314.1 | - | 252 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |