Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097123.4 | XP_001097123.2 | 740 | 86 | ||
GenBank mRNA | JV050388.1 | - | 228 | 86 | ||
JV640955.1 | - | 213 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |