Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047146.2 | NP_001040611.2 | 134 | 61 | ||
GenBank mRNA | CN644864.1 | - | 134 | 61 |
Gene Symbol: HBG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833778.1 | XP_028689611.1 | 492 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |