Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112611.4 | XP_001112611.1 | 119 | 117 | ||
XM_015132873.2 | XP_014988359.1 | 119 | 117 | |||
XM_028837278.1 | XP_028693111.1 | 119 | 117 | |||
GenBank mRNA | JU496638.1 | - | 134 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |