Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266020.1 | NP_001252949.1 | 155 | 108 | ||
GenBank mRNA | JU473468.1 | - | 155 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta Inventoried |