Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205127.2 | NP_001192056.2 | 3544 | 105 | ||
XM_015112348.2 | XP_014967834.1 | 3103 | 105 | |||
XM_015112349.2 | XP_014967835.2 | 2405 | 105 | |||
XM_015112350.2 | XP_014967836.1 | 941 | 105 | |||
GenBank mRNA | CK231727.1 | - | 372 | 105 | ||
JU471546.1 | - | 759 | 105 | |||
JV045048.1 | - | 653 | 105 | |||
JV588048.1 | - | 3761 | 105 | |||
JV622917.1 | - | 3441 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |