Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121664.2 | XP_014977150.2 | 789 | 137 | ||
XM_015121665.2 | XP_014977151.2 | 789 | 137 | |||
XM_028837909.1 | XP_028693742.1 | 753 | 137 | |||
XM_028837911.1 | XP_028693744.1 | 711 | 137 | |||
XM_028837912.1 | XP_028693745.1 | 660 | 137 | |||
XM_028837913.1 | XP_028693746.1 | 894 | 137 | |||
XM_028837914.1 | XP_028693747.1 | 779 | 137 | |||
GenBank mRNA | JV663985.1 | - | 767 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |