Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278397.1 | NP_001265326.1 | 593 | 105 | ||
XM_015115026.2 | XP_014970512.2 | 469 | 105 | |||
XM_015115027.2 | XP_014970513.2 | 468 | 105 | |||
XM_015115028.2 | XP_014970514.2 | 476 | 105 | |||
XM_015115029.2 | XP_014970515.2 | 469 | 105 | |||
XM_015115031.2 | XP_014970517.2 | 466 | 105 | |||
XM_015115032.2 | XP_014970518.2 | 487 | 105 | |||
XM_015115033.2 | XP_014970519.2 | 600 | 105 | |||
XM_028832496.1 | XP_028688329.1 | 892 | 105 | |||
XM_028832498.1 | XP_028688331.1 | 473 | 105 | |||
GenBank mRNA | AJ294467.1 | - | 137 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |