Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130424.1 | NP_001123896.1 | 2257 | 79 | ||
XM_028847926.1 | XP_028703759.1 | 3562 | 79 | |||
XM_028847927.1 | XP_028703760.1 | 3470 | 79 | |||
GenBank mRNA | AB445623.1 | - | 2257 | 79 | ||
AY525611.1 | - | 352 | 79 | |||
EU204930.1 | - | 2288 | 79 | |||
JU329634.1 | - | 910 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |