Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032955.1 | NP_001028127.1 | 379 | 59 | ||
GenBank mRNA | AY574369.1 | - | 379 | 59 | ||
JV650519.1 | - | 415 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta Inventoried |