Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090729.4 | XP_001090729.4 | 946 | 78 | ||
GenBank mRNA | AY574371.1 | - | 131 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta Inventoried |