Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037862.1 | NP_001032951.1 | 699 | 92 | ||
GenBank mRNA | AB180075.1 | - | 699 | 92 | ||
JV722023.1 | - | 56 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Macaca mulatta |