Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266124.1 | NP_001253053.1 | 506 | 65 | ||
GenBank mRNA | AF252388.1 | - | 303 | 65 | ||
JU326959.1 | - | 307 | 65 | |||
JU473867.1 | - | 483 | 65 | |||
JV045638.1 | - | 506 | 65 | |||
JV637471.1 | - | 499 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |