Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032962.1 | NP_001028134.1 | 488 | 80 | ||
GenBank mRNA | AY321585.1 | - | 488 | 80 | ||
JU473796.1 | - | 455 | 80 | |||
JV043495.1 | - | 637 | 80 | |||
JV635753.1 | - | 771 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Macaca mulatta |