Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032930.1 | NP_001028102.1 | 1949 | 69 | ||
GenBank mRNA | AY340665.1 | - | 1949 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |