Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047137.1 | NP_001040602.1 | 83 | 62 | ||
XM_015142098.2 | XP_014997584.1 | 158 | 62 | |||
GenBank mRNA | AY349163.1 | - | 83 | 62 | ||
AY445834.1 | - | 83 | 62 | |||
JV045380.1 | - | 81 | 62 | |||
JV669635.1 | - | 136 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |