Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039950.1 | NP_001035039.1 | 653 | 65 | ||
XM_015124857.2 | XP_014980343.2 | 503 | 65 | |||
GenBank mRNA | AY386684.1 | - | 653 | 65 | ||
AY386686.1 | - | 326 | 65 | |||
AY386687.1 | - | 362 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |