Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195473.1 | NP_001182402.1 | 405 | 119 | ||
GenBank mRNA | AY445037.2 | - | 355 | 119 | ||
CO648448.1 | - | 417 | 119 | |||
JU477783.1 | - | 420 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |