Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037284.1 | NP_001032361.1 | 665 | 80 | ||
GenBank mRNA | AY680445.1 | - | 186 | 80 | ||
CN641767.1 | - | 665 | 80 | |||
JU322674.1 | - | 698 | 80 | |||
JU332303.1 | - | 698 | 80 | |||
JU470877.1 | - | 699 | 80 | |||
JV048021.1 | - | 709 | 80 |
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV715946.1 | - | 5475 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |