Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118620.2 | XP_014974106.1 | 500 | 71 | ||
GenBank mRNA | AY680486.1 | - | 145 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |