Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02788868_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
EGK_12829, GPR56
Chromosome Location:
Chr.20: 44855566 - 44904737 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02788868_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001033020.1
NP_001028192.1
938
76
XM_015126156.2
XP_014981642.1
1086
76
XM_015126157.2
XP_014981643.1
1232
76
XM_015126158.2
XP_014981644.1
1044
76
XM_015126161.2
XP_014981647.1
1044
76
XM_015126162.2
XP_014981648.1
1062
76
XM_015126163.2
XP_014981649.1
1114
76
XM_015126164.2
XP_014981650.1
1083
76
XM_015126165.2
XP_014981651.1
1113
76
XM_015126166.2
XP_014981652.1
1282
76
XM_015126167.2
XP_014981653.1
1029
76
XM_015126168.2
XP_014981654.1
1074
76
XM_015126169.2
XP_014981655.1
1095
76
XM_015126170.2
XP_014981656.1
1046
76
XM_015126171.2
XP_014981657.1
1531
76
XM_015126172.2
XP_014981658.1
1516
76
XM_028840341.1
XP_028696174.1
977
76
XM_028840342.1
XP_028696175.1
974
76
XM_028840344.1
XP_028696177.1
1179
76
XM_028840345.1
XP_028696178.1
961
76
XM_028840346.1
XP_028696179.1
986
76
XM_028840347.1
XP_028696180.1
1036
76
GenBank mRNA
AY845352.1
-
938
76
JU323293.1
-
1172
76
JU337110.1
-
1172
76
JU470897.1
-
1182
76
JU470898.1
-
1167
76
JU477855.1
-
1167
76
JV601061.1
-
1172
76
JV635411.1
-
1146
76
Back To Top
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership:
Amplicon length between 71 and 85
G-protein coupled receptor
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0