Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040415.2 | NP_001035505.1 | 588 | 101 | ||
GenBank mRNA | AY913830.1 | - | 590 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |