Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261815.2 | NP_001248744.2 | 1179 | 94 | ||
XM_028844747.1 | XP_028700580.1 | 1811 | 94 | |||
XM_028844748.1 | XP_028700581.1 | 1812 | 94 | |||
XM_028844749.1 | XP_028700582.1 | 1806 | 94 | |||
XM_028844750.1 | XP_028700583.1 | 1812 | 94 | |||
XM_028844751.1 | XP_028700584.1 | 1742 | 94 | |||
XM_028844752.1 | XP_028700585.1 | 1418 | 94 | |||
XM_028844753.1 | XP_028700586.1 | 1435 | 94 | |||
XM_028844754.1 | XP_028700587.1 | 1810 | 94 | |||
XM_028844755.1 | XP_028700588.1 | 1418 | 94 | |||
XM_028844756.1 | XP_028700589.1 | 1593 | 94 | |||
XM_028844757.1 | XP_028700590.1 | 1203 | 94 | |||
XM_028844758.1 | XP_028700591.1 | 1418 | 94 | |||
XM_028844759.1 | XP_028700592.1 | 1418 | 94 | |||
GenBank mRNA | AY950560.1 | - | 143 | 94 | ||
JU327588.1 | - | 1150 | 94 | |||
JU327589.1 | - | 1144 | 94 | |||
JU471703.1 | - | 1183 | 94 | |||
JU474451.1 | - | 1183 | 94 | |||
JV566287.1 | - | 1224 | 94 | |||
JV635975.1 | - | 1243 | 94 | |||
JV635976.1 | - | 1237 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |