Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195660.1 | NP_001182589.1 | 457 | 100 | ||
GenBank mRNA | CB308818.1 | - | 460 | 100 | ||
CO644659.1 | - | 443 | 100 | |||
DQ147924.1 | - | 360 | 101 | |||
JU326829.1 | - | 335 | 100 | |||
JU472778.1 | - | 445 | 100 | |||
JV045909.1 | - | 461 | 100 | |||
JV635773.1 | - | 469 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 3' Most Macaca mulatta |