Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195652.1 | NP_001182581.1 | 1141 | 80 | ||
GenBank mRNA | CO647004.1 | - | 511 | 80 | ||
DQ148152.1 | - | 60 | 80 | |||
JU326689.1 | - | 1103 | 80 | |||
JU476699.1 | - | 1193 | 80 | |||
JV044390.1 | - | 1196 | 80 | |||
JV590057.1 | - | 1163 | 80 | |||
JV658162.1 | - | 1194 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |