Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038662.1 | NP_001033751.1 | 215 | 130 | ||
XM_028838330.1 | XP_028694163.1 | 369 | 130 | |||
XM_028838331.1 | XP_028694164.1 | 369 | 130 | |||
XM_028838333.1 | XP_028694166.1 | 372 | 130 | |||
GenBank mRNA | DQ227271.1 | - | 215 | 130 |
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123131.2 | XP_014978617.1 | 573 | 130 | ||
XM_015123132.2 | XP_014978618.1 | 573 | 130 | |||
XM_015123133.2 | XP_014978619.1 | 572 | 130 | |||
XM_015123134.2 | XP_014978620.1 | 573 | 130 | |||
XM_028839871.1 | XP_028695704.1 | 573 | 130 | |||
XM_028839872.1 | XP_028695705.1 | 573 | 130 | |||
XM_028839873.1 | XP_028695706.1 | 407 | 130 | |||
GenBank mRNA | JU478010.1 | - | 290 | 130 | ||
JU478011.1 | - | 290 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Macaca mulatta |