Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847001.1 | XP_028702834.1 | 4062 | 91 | ||
GenBank mRNA | J04635.1 | - | 2148 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |