Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044725.1 | NP_001038190.1 | 859 | 82 | ||
XM_028849042.1 | XP_028704875.1 | 1194 | 82 | |||
GenBank mRNA | JU485898.1 | - | 145 | 82 | ||
U19841.1 | - | 859 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |