Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047130.1 | NP_001040595.1 | 359 | 146 | ||
GenBank mRNA | U19847.1 | - | 359 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta Inventoried |