Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048246.2 | NP_001041711.2 | 1082 | 159 | ||
GenBank mRNA | AF157396.1 | - | 1080 | 159 | ||
EF580137.1 | - | 1086 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |