Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF190979.1 | - | 91 | 154 | ||
AF190981.1 | - | 91 | 154 | |||
AF190982.1 | - | 91 | 154 | |||
AF190984.1 | - | 91 | 154 | |||
AF395620.1 | - | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |