Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF580143.1 | - | 1123 | 100 | ||
U41379.1 | - | 1088 | 100 |
Gene Symbol: LOC720132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846911.1 | XP_028702744.1 | 1265 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |