Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032954.1 | NP_001028126.1 | 663 | 108 | ||
GenBank mRNA | AY462045.1 | - | 663 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |