Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093817.3 | XP_001093817.1 | 1375 | 117 | ||
XM_001094648.4 | XP_001094648.1 | 1381 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |