Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115817.2 | XP_014971303.1 | 2062 | 143 | ||
XM_015115818.2 | XP_014971304.1 | 2047 | 143 | |||
GenBank mRNA | JV726677.1 | - | 257 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |