Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261663.1 | NP_001248592.1 | 409 | 121 | ||
GenBank mRNA | JU333391.1 | - | 464 | 121 | ||
JU472949.1 | - | 393 | 121 | |||
JV046449.1 | - | 409 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |