Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261520.1 | NP_001248449.1 | 137 | 157 | ||
GenBank mRNA | JU334917.1 | - | 131 | 157 | ||
JU475925.1 | - | 136 | 157 | |||
JV047321.1 | - | 137 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |