Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266625.1 | NP_001253554.1 | 429 | 89 | ||
XM_015115449.2 | XP_014970935.1 | 456 | 89 | |||
GenBank mRNA | JU328134.1 | - | 317 | 89 | ||
JU475916.1 | - | 335 | 89 | |||
JV043389.1 | - | 429 | 89 |