Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261020.1 | NP_001247949.1 | 353 | 94 | ||
XM_015134600.2 | XP_014990086.1 | 669 | 94 | |||
XM_015134601.2 | XP_014990087.1 | 273 | 94 | |||
XM_015134602.2 | XP_014990088.1 | 609 | 94 | |||
XM_015134607.2 | XP_014990093.1 | 845 | 94 | |||
XM_015134609.2 | XP_014990095.1 | 756 | 94 | |||
XM_015134610.2 | XP_014990096.1 | 512 | 94 | |||
XM_028844848.1 | XP_028700681.1 | 1054 | 94 | |||
GenBank mRNA | JU322151.1 | - | 352 | 94 | ||
JV046752.1 | - | 353 | 94 |
Gene Symbol: COG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU478818.1 | - | 5922 | 94 | ||
JV705247.1 | - | 5901 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |