Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095770.4 | XP_001095770.1 | 529 | 105 | ||
XM_015134298.2 | XP_014989784.1 | 585 | 105 | |||
GenBank mRNA | JU325900.1 | - | 452 | 105 | ||
JV063770.1 | - | 495 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |