Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104583.1 | NP_001098053.1 | 673 | 81 | ||
GenBank mRNA | EF057492.1 | - | 673 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |