Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266092.2 | NP_001253021.1 | 131 | 150 | ||
GenBank mRNA | JU332582.1 | - | 122 | 150 | ||
JU474049.1 | - | 124 | 150 | |||
JV045969.1 | - | 134 | 150 | |||
JV640527.1 | - | 131 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |