Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266092.2 | NP_001253021.1 | 582 | 106 | ||
GenBank mRNA | JU332582.1 | - | 573 | 106 | ||
JU474049.1 | - | 575 | 106 | |||
JV045969.1 | - | 585 | 106 | |||
JV640527.1 | - | 582 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |