Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261171.1 | NP_001248100.1 | 607 | 114 | ||
GenBank mRNA | JU322857.1 | - | 607 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |