Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260538.1 | NP_001247467.1 | 652 | 148 | ||
GenBank mRNA | JV046098.1 | - | 652 | 148 |
Gene Symbol: SUPT20H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335347.1 | - | 3120 | 148 | ||
JU335348.1 | - | 3016 | 148 | |||
JU476308.1 | - | 3078 | 148 | |||
JU476309.1 | - | 2974 | 148 | |||
JV718351.1 | - | 3150 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |