Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082556.4 | XP_001082556.1 | 272 | 64 | ||
XM_002806242.3 | XP_002806288.1 | 224 | 64 | |||
XM_028842087.1 | XP_028697920.1 | 287 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |