Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260894.1 | NP_001247823.1 | 373 | 115 | ||
XM_015149346.2 | XP_015004832.1 | 642 | 115 | |||
GenBank mRNA | JU321746.1 | - | 275 | 115 | ||
JU473447.1 | - | 323 | 115 | |||
JV044849.1 | - | 373 | 115 | |||
JV452520.1 | - | 322 | 115 | |||
JV490286.1 | - | 335 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |