Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260608.1 | NP_001247537.1 | 341 | 133 | ||
XM_028849098.1 | XP_028704931.1 | 194 | 133 | |||
GenBank mRNA | JU321610.1 | - | 407 | 133 | ||
JV045144.1 | - | 341 | 133 | |||
JV601372.1 | - | 349 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |