Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193692.1 | NP_001180621.1 | 471 | 100 | ||
XM_015114004.2 | XP_014969490.1 | 635 | 100 | |||
XM_015114005.2 | XP_014969491.1 | 433 | 100 | |||
XM_028832505.1 | XP_028688338.1 | 492 | 100 | |||
GenBank mRNA | JU321399.1 | - | 401 | 100 | ||
JU473320.1 | - | 403 | 100 | |||
JV047471.1 | - | 406 | 100 | |||
JV472769.1 | - | 400 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |